Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cltb
Entrez Gene ID
74325
Gene Name
clathrin, light polypeptide (Lcb)
Gene Aliases
2310046E19Rik
Chromosome Location
Chr.13: 54592939 - 54611397 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC070404.1 - 4 - 5 478 75

View Full Product Details