Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnot10
Entrez Gene ID
78893
Gene Name
CCR4-NOT transcription complex, subunit 10
Gene Aliases
2600001P13Rik
Chromosome Location
Chr.9: 114585874 - 114640216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153585.5 NP_705813.2 19 - 20 2516 71
GenBank mRNA AK029103.1 - 19 - 20 2391 71
BC025554.1 - 19 - 20 2430 71
BC036178.1 - 16 - 17 1972 71
BC039183.1 - 19 - 20 2490 71
BC047204.1 - 3 - 4 528 71
CX222096.1 - 1 - 2 105 71

View Full Product Details