Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iffo2
Entrez Gene ID
212632
Gene Name
intermediate filament family orphan 2
Gene Aliases
C79267
Chromosome Location
Chr.4: 139530548 - 139620382 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205173.1 NP_001192102.1 3 - 4 1035 82
NM_183148.3 NP_898971.1 6 - 7 655 82
GenBank mRNA BC027170.1 - 2 - 3 181 82
BC100381.1 - 6 - 7 655 82

View Full Product Details