Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tspan6
Entrez Gene ID
56496
Gene Name
tetraspanin 6
Gene Aliases
6720473L21Rik, AI316786, T245, TSPAN-6, Tm4sf, Tm4sf6
Chromosome Location
Chr.X: 133891070 - 133898429 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019656.3 NP_062630.2 2 - 3 414 98
GenBank mRNA AF053454.1 - 2 - 3 361 98
AK146673.1 - 2 - 3 319 98
AK149204.1 - 2 - 3 324 98
AK160313.1 - 2 - 3 308 98
AK167384.1 - 2 - 3 310 98
BC003733.1 - 2 - 3 348 98
BC110314.1 - 2 - 3 323 98

View Full Product Details