Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dnm3
- Entrez Gene ID
-
103967
- Gene Name
- dynamin 3
- Gene Aliases
-
9630020E24Rik, AW061159, B230343F03Rik, C530045C17, mKIAA0820
- Chromosome Location
-
Chr.1: 161982450 - 162478323 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC004050.1 |
- |
8 - 9 |
1084 |
85 |
View Full Product Details