Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Agrn
Entrez Gene ID
11603
Gene Name
agrin
Gene Aliases
Agrin, nmf380
Chromosome Location
Chr.4: 156165290 - 156197519 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021604.3 NP_067617.3 5 - 6 1020 84
GenBank mRNA AK147577.1 - 4 - 5 1041 84
BC058651.1 - 4 - 5 1133 84
BC150703.1 - 3 - 4 474 84

View Full Product Details