The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008112.4 | NP_032138.3 | 9 - 10 | 1364 | 100 |
GenBank mRNA | AK007871.1 | - | 9 - 10 | 1307 | 100 |
AK078144.1 | - | - | 1307 | 100 | |
AK079690.1 | - | 8 - 9 | 1205 | 100 | |
AK090083.1 | - | 9 - 10 | 1312 | 100 | |
AK137761.1 | - | 9 - 10 | 1311 | 100 | |
AK143633.1 | - | 9 - 10 | 1309 | 100 | |
AK143681.1 | - | 9 - 10 | 1343 | 100 | |
AK149958.1 | - | 9 - 10 | 1317 | 100 | |
AK150356.1 | - | 9 - 10 | 1299 | 100 | |
AK150655.1 | - | 9 - 10 | 1334 | 100 | |
AK152250.1 | - | 9 - 10 | 1298 | 100 | |
AK153381.1 | - | 9 - 10 | 1277 | 100 | |
AK159734.1 | - | 9 - 10 | 1309 | 100 | |
AK166317.1 | - | 9 - 10 | 1309 | 100 | |
AK167659.1 | - | 9 - 10 | 1321 | 100 | |
AK169299.1 | - | 9 - 10 | 1275 | 100 | |
BC024971.1 | - | 9 - 10 | 1285 | 100 | |
BC055341.1 | - | 9 - 10 | 1290 | 100 |