Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdv3
Entrez Gene ID
321022
Gene Name
carnitine deficiency-associated gene expressed in ventricle 3
Gene Aliases
2510010F10Rik, C230084J24Rik, C79446, TPP36
Chromosome Location
Chr.9: 103353102 - 103365835 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134427.1 NP_001127899.1 4 - 5 892 91
NM_175565.3 NP_780774.1 4 - 5 1102 91
GenBank mRNA AF215660.1 - 4 - 5 860 91
AK172053.1 - 4 - 5 811 91

View Full Product Details