Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2b23
Entrez Gene ID
243881
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 23
Gene Aliases
EG243881
Chromosome Location
Chr.7: 26665227 - 26690372 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081148.1 NP_001074617.1 4 - 5 646 100

View Full Product Details