Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Serpina3i
- Entrez Gene ID
-
628900
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade A, member 3I
- Gene Aliases
-
2B2, EG628900, Gm6930
- Chromosome Location
-
Chr.12: 104263045 - 104269365 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK019935.1 |
- |
1 - 2 |
59 |
105 |
View Full Product Details