Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mup21
Entrez Gene ID
381531
Gene Name
major urinary protein 21
Gene Aliases
Gm11208, Mup, Mup26, OTTMUSG00000000231, bM64F17.1, bM64F17.4
Chromosome Location
Chr.4: 62147832 - 62150841 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009550.2 NP_001009550.1 3 - 4 395 155
GenBank mRNA BC141241.1 - 3 - 4 390 155
EU882236.1 - 3 - 4 306 155

View Full Product Details