Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adam20
Entrez Gene ID
384806
Gene Name
a disintegrin and metallopeptidase domain 20
Gene Aliases
4930529F22Rik, Adam38
Chromosome Location
Chr.8: 40793273 - 40797342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009548.2 NP_001009548.1 2 - 2 2406 137
GenBank mRNA AK133113.1 - 2 - 2 2407 137
BC150793.1 - - 2292 137

View Full Product Details