Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rpl36al
- Entrez Gene ID
-
66483
- Gene Name
- ribosomal protein L36A-like
- Gene Aliases
-
2410038A03Rik, RPL36A
- Chromosome Location
-
Chr.12: 69182734 - 69184067 on Build GRCm38
- Gene Symbol
- Dnaaf2
- Entrez Gene ID
-
109065
- Gene Name
- dynein, axonemal assembly factor 2
- Gene Aliases
-
1110034A24Rik, 2810020C19Rik, AV032410, Ktu, TU54, kintoun, mknt
- Chromosome Location
-
Chr.12: 69189838 - 69198429 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EU599049.1 |
- |
- |
1002 |
99 |
View Full Product Details