Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgfbp3
Entrez Gene ID
72514
Gene Name
fibroblast growth factor binding protein 3
Gene Aliases
2610306H15Rik, Fgf-bp3
Chromosome Location
Chr.19: 36917550 - 36919599 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028263.1 NP_082539.2 1 - 2 32 56
GenBank mRNA AK167949.1 - 1 - 2 50 56
BC055778.1 - 1 - 2 32 56

View Full Product Details