The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028660.3 | NP_082936.2 | 2 - 3 | 310 | 70 |
GenBank mRNA | AK004807.1 | - | 2 - 3 | 241 | 70 |
BC109326.1 | - | 2 - 3 | 222 | 70 | |
BC109327.1 | - | 2 - 3 | 222 | 70 | |
BX520246.1 | - | 2 - 3 | 310 | 70 |