Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc9
Entrez Gene ID
320116
Gene Name
fibronectin type III domain containing 9
Gene Aliases
C030019I05Rik
Chromosome Location
Chr.11: 46235557 - 46239871 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177075.4 NP_796049.1 1 - 2 37 64
GenBank mRNA AK081240.1 - 1 - 2 38 64
BY728197.1 - 1 - 2 38 64

View Full Product Details