The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010732.4 | NP_034862.1 | 1 - 2 | 326 | 64 |
GenBank mRNA | AK036271.1 | - | 1 - 2 | 327 | 64 |
AK039216.1 | - | 1 - 2 | 327 | 64 | |
BC056458.1 | - | 1 - 2 | 68 | 64 |