Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Znrf4
Entrez Gene ID
20834
Gene Name
zinc and ring finger 4
Gene Aliases
1700094M01Rik, Ssrzf1, spzn
Chromosome Location
Chr.17: 56511248 - 56512483 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK007069.1 - - 271 133

View Full Product Details