The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289760.1 | NP_001276689.1 | 2 - 3 | 662 | 74 |
GenBank mRNA | AK190447.1 | - | 1 - 2 | 155 | 74 |
X56574.1 | - | 1 - 2 | 452 | 74 |