The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020608.3 | NP_065633.2 | 10 - 11 | 2385 | 65 |
GenBank mRNA | AK129356.1 | - | 1 - 2 | 561 | 65 |
AK147411.1 | - | 10 - 11 | 2386 | 65 | |
AK147449.1 | - | 10 - 11 | 2386 | 65 |