Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hottip
- Entrez Gene ID
-
791364
- Gene Name
- Hoxa distal transcript antisense RNA
- Gene Aliases
-
ENSMUSG00000055408, Gm15053, OTTMUSG00000018874
- Chromosome Location
-
Chr.6: 52262775 - 52267603 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110441.1 |
|
1 - 2 |
645 |
132 |
View Full Product Details