Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ercc8
- Entrez Gene ID
-
71991
- Gene Name
- excision repaiross-complementing rodent repair deficiency, complementation group 8
- Gene Aliases
-
2410022P04Rik, 2810431L23Rik, 4631412O06Rik, B130065P18Rik, Ckn1, Csa
- Chromosome Location
-
Chr.13: 108158694 - 108199284 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK013216.1 |
- |
5 - 6 |
542 |
175 |
View Full Product Details