The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033596.3 | NP_291074.1 | 1 - 1 | 147 | 179 |
GenBank mRNA | BC051566.1 | - | 1 - 1 | 155 | 179 |
BC057955.1 | - | - | 141 | 179 | |
BX520915.1 | - | 1 - 1 | 147 | 179 |