The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024413.2 | NP_077724.2 | 2 - 2 | 168 | 180 |
GenBank mRNA | AK143976.1 | - | - | 157 | 180 |
AK171398.1 | - | 2 - 2 | 157 | 180 | |
AW227287.1 | - | 2 - 2 | 168 | 180 |