Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BC100530
- Entrez Gene ID
-
100034684
- Gene Name
- cDNA sequence BC100530
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 36359382 - 36367570 on Build GRCm38
- Gene Symbol
- BC117090
- Entrez Gene ID
-
100038854
- Gene Name
- cDNA sequence BC1179090
- Gene Aliases
-
Gm1975
- Chromosome Location
-
Chr.16: 36321665 - 36334332 on Build GRCm38
- Gene Symbol
- Stfa1
- Entrez Gene ID
-
20861
- Gene Name
- stefin A1
- Gene Aliases
-
Stefin-3, Stf-3, Stf1, Stf3, Stfa3
- Chromosome Location
-
Chr.16: 36277148 - 36285371 on Build GRCm38
View Full Product Details