Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P2ry14
Entrez Gene ID
140795
Gene Name
purinergic receptor P2Y, G-protein coupled, 14
Gene Aliases
A330108O13Rik, Gpr105, P2Y14
Chromosome Location
Chr.3: 59114623 - 59153492 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008497.2 NP_001008497.1 3 - 3 1592 157
NM_001287119.1 NP_001274048.1 3 - 3 1397 157
NM_001287120.1 NP_001274049.1 2 - 2 1522 157
NM_001287121.1 NP_001274050.1 3 - 3 1648 157
NM_001287122.1 NP_001274051.1 2 - 2 1725 157
NM_001287123.1 NP_001274052.1 2 - 2 1774 157
NM_001287124.1 NP_001274053.1 2 - 2 1488 157
NM_133200.4 NP_573463.1 2 - 2 1718 157
GenBank mRNA AK140301.1 - 2 - 2 1550 157
AK155035.1 - 3 - 3 1401 157
AK164982.1 - 2 - 2 1537 157
BC028995.1 - 2 - 2 1527 157
BC058558.1 - 3 - 3 1454 157

View Full Product Details