The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009548.2 | NP_001009548.1 | 2 - 2 | 2377 | 132 |
GenBank mRNA | AK133113.1 | - | 2 - 2 | 2378 | 132 |
BC150793.1 | - | - | 2263 | 132 |