Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt77
Entrez Gene ID
406220
Gene Name
keratin 77
Gene Aliases
4732484G22Rik, K1emb, Krt239, mK1b
Chromosome Location
Chr.15: 101858731 - 101869706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003667.1 NP_001003667.1 2 - 3 768 153
GenBank mRNA AK132513.1 - 2 - 3 856 153
AY379112.1 - 1 - 2 207 153
AY651826.1 - 1 - 2 207 153
BC107395.2 - 2 - 3 768 153
BC107396.2 - 2 - 3 768 153
BC141259.1 - 2 - 3 791 153

View Full Product Details