Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt77
Entrez Gene ID
406220
Gene Name
keratin 77
Gene Aliases
4732484G22Rik, K1emb, Krt239, mK1b
Chromosome Location
Chr.15: 101858731 - 101869706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003667.1 NP_001003667.1 3 - 4 833 145
GenBank mRNA AK132513.1 - 3 - 4 921 145
AY379112.1 - 2 - 3 272 145
AY651826.1 - 2 - 3 272 145
BC107395.2 - 3 - 4 833 145
BC107396.2 - 3 - 4 833 145
BC141259.1 - 3 - 4 856 145

View Full Product Details