The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_205844.3 | NP_995316.2 | 5 - 6 | 898 | 121 |
GenBank mRNA | AY457637.2 | - | 5 - 6 | 898 | 121 |