The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012520.2 | NP_001012538.2 | 3 - 4 | 393 | 75 |
GenBank mRNA | AY324874.1 | - | 3 - 4 | 393 | 75 |