The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007420.3 | NP_031446.2 | 1 - 1 | 1977 | 75 |
GenBank mRNA | AK080276.1 | - | 1 - 1 | 1978 | 75 |
BC030346.1 | - | 1 - 1 | 1429 | 75 | |
BC032883.1 | - | 1 - 1 | 1955 | 75 |