Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nhp2l1
Entrez Gene ID
20826
Gene Name
NHP2 non-histone chromosome protein 2-like 1 (S. cerevisiae)
Gene Aliases
FA-1, Fta1, Ssfa1
Chromosome Location
Chr.15: 82041345 - 82047598 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA U95114.1 - 3 - 3 260 68

View Full Product Details