The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013761.2 | NP_001013783.1 | 3 - 4 | 483 | 65 |
NM_001285916.1 | NP_001272845.1 | 2 - 3 | 577 | 65 | |
GenBank mRNA | AK141059.1 | - | 2 - 3 | 577 | 65 |
BC086669.1 | - | 2 - 3 | 238 | 65 |