Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim80
Entrez Gene ID
432613
Gene Name
tripartite motif-containing 80
Gene Aliases
4933422H20Rik, Trim47l
Chromosome Location
Chr.11: 115440545 - 115448268 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033775.3 NP_001028947.1 2 - 3 109 70
GenBank mRNA AK133415.1 - 2 - 3 110 70
BC147608.1 - 2 - 3 127 70
BC157912.1 - 2 - 3 127 70

View Full Product Details