Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc13
Entrez Gene ID
100502861
Gene Name
coiled-coil domain containing 13
Gene Aliases
2900041A11Rik, Gm188
Chromosome Location
Chr.9: 121797624 - 121839515 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK158659.1 - 5 - 6 661 93
BC147354.1 - 5 - 6 626 93

View Full Product Details