The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009081.2 | NP_033107.1 | 3 - 4 | 248 | 114 |
GenBank mRNA | AK012368.1 | - | 3 - 4 | 249 | 114 |
AK018714.1 | - | 3 - 4 | 249 | 114 | |
AK076133.1 | - | 3 - 4 | 249 | 114 | |
BC024395.1 | - | 3 - 4 | 248 | 114 | |
BC086797.1 | - | 3 - 4 | 234 | 114 | |
BC086932.1 | - | 3 - 4 | 250 | 114 | |
BC092012.1 | - | 3 - 4 | 248 | 114 | |
X74856.1 | - | 3 - 4 | 276 | 114 |