Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Defb44-ps
- Entrez Gene ID
-
654454
- Gene Name
- defensin beta 44, pseudogene
- Gene Aliases
-
Defb44, Defb49, Defb49-ps, EG574082
- Chromosome Location
-
Chr.1: 18210053 - 18223564 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002879.2 |
|
2 - 2 |
131 |
103 |
GenBank mRNA |
AY621334.1 |
- |
2 - 2 |
130 |
104 |
DQ012049.1 |
- |
2 - 2 |
|
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cysteine-rich secretory protein 4 |
Crisp4 |
9230112K08Rik |
-64.1kb
|
Sense |
Chr. - 18114905 - 18145967 on Build GRCm38 |
Downstream |
defensin beta 18 |
Defb18 |
BD-18, EG654460, mBD-18 |
26.2kb
|
Sense |
Chr. - 18236473 - 18237443 on Build GRCm38 |
View Full Product Details