Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekhm1
Entrez Gene ID
353047
Gene Name
pleckstrin homology domain containing, family M (with RUN domain) member 1
Gene Aliases
AP162, B2, BC038943, D330036J23Rik
Chromosome Location
Chr.11: 103365092 - 103412664 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183034.1 NP_898855.1 6 - 7 1746 94
GenBank mRNA AK154703.1 - 5 - 6 1675 94
AK155357.1 - 6 - 7 1770 94
AK157752.1 - 5 - 6 1390 94
BC053079.1 - 6 - 7 1746 94

View Full Product Details