The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113470.1 | NP_001106941.1 | 6 - 7 | 1025 | 61 |
NM_146012.2 | NP_666124.1 | 1 - 2 | 338 | 61 | |
GenBank mRNA | AK045068.1 | - | 6 - 7 | 781 | 61 |
AK049271.1 | - | 6 - 7 | 825 | 61 | |
AK170287.1 | - | 6 - 7 | 726 | 61 | |
BC025650.1 | - | 1 - 2 | 350 | 61 | |
BC085142.1 | - | 6 - 7 | 1025 | 61 | |
BC089307.1 | - | 5 - 6 | 330 | 61 | |
CB527995.1 | - | 1 - 2 | 328 | 61 |