Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Drg2
Entrez Gene ID
13495
Gene Name
developmentally regulated GTP binding protein 2
Gene Aliases
AI255295, AI461605
Chromosome Location
Chr.11: 60454617 - 60468724 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021354.1 NP_067329.1 3 - 4 382 114
GenBank mRNA AJ243590.1 - 3 - 4 382 114
BC082564.1 - 3 - 4 338 114

View Full Product Details