The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009370.2 | NP_033396.1 | 9 - 9 | 5521 | 131 |
GenBank mRNA | AK154381.1 | - | 8 - 8 | 5284 | 131 |
BC063260.1 | - | 9 - 9 | 5521 | 131 |