The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164789.1 | NP_001158261.1 | 6 - 6 | 3551 | 149 |
NM_010136.3 | NP_034266.2 | 6 - 6 | 3608 | 149 | |
GenBank mRNA | AK081881.1 | - | - | 1002 | 149 |
BC094319.1 | - | 6 - 6 | 3454 | 150 |