The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011356.4 | NP_035486.1 | 6 - 6 | 2843 | 66 |
GenBank mRNA | AK029941.1 | - | 6 - 6 | 2845 | 66 |
BC016884.1 | - | 6 - 6 | 2686 | 66 |