Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps4x
Entrez Gene ID
20102
Gene Name
ribosomal protein S4, X-linked
Gene Aliases
Rps4, Rps4-1
Chromosome Location
Chr.X: 102184941 - 102189306 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009094.1 NP_033120.1 4 - 5 547 127
GenBank mRNA AK002445.1 - 5 - 6 565 127
AK010624.1 - 5 - 6 568 127
AK010676.1 - 5 - 6 565 127
AK010921.1 - 5 - 6 565 127
AK012429.1 - 5 - 6 565 127
AK014210.1 - 5 - 6 563 127
AK088410.1 - 5 - 6 565 127
AK135369.1 - 5 - 6 568 127
AK153211.1 - 5 - 6 559 127
BC009100.1 - 4 - 5 572 127
BC089348.1 - 4 - 5 549 127
BC089349.1 - 4 - 5 543 127
BC106152.1 - 4 - 5 548 127
M73436.1 - 4 - 5 547 127

View Full Product Details