The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NR_003188.1 | 1 - 1 | 1734 | 123 | ||
GenBank mRNA | AK084356.1 | - | 2 - 2 | 1166 | 123 | |
AY940614.1 | - | LIT3387 Kis2 |
1 - 1 | 1734 | 123 | |
AY940615.1 | - | LIT3388 Kis2 |
2 - 2 | 1420 | 123 | |
AY940616.1 | - | LIT3389 Kis2 |
1 - 1 | 1053 | 123 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | microRNA 18b | Mir18b | Mirn18b, mir-18b, mmu-mir-18b | -150.0 | Sense | Chr. - 52742331 - 52742413 on Build GRCm38 |
Downstream | coiled-coil domain containing 160 | Ccdc160 | 6330534C20Rik | 47.3kb | Anti-sense | Chr. - 52791200 - 52799474 on Build GRCm38 |