The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135991.1 | NP_001129463.1 | 1 - 1 | 856 | 75 |
GenBank mRNA | AK020700.1 | - | 1 - 1 | 884 | 75 |
BC115499.1 | - | 1 - 1 | 856 | 75 | |
BC115500.1 | - | 1 - 1 | 856 | 75 |