Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snhg7
Entrez Gene ID
72091
Gene Name
small nucleolar RNA host gene 7
Gene Aliases
2610002F03Rik, AI427062
Chromosome Location
Chr.2: 26637176 - 26640244 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024068.2 2 - 3 230 66
GenBank mRNA AK011283.1 - 2 - 3 231 66
AK028166.1 - 2 - 3 231 66


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY710585.1 - 2 - 3 227 Chr. 2 - 26637229 - 26640241 [-] on Build GRCm38 66
GenBankmRNA BY710095.1 - 2 - 3 225 Chr. 2 - 26637299 - 26640239 [-] on Build GRCm38 66
GenBankmRNA EG359318.1 - Chr. - - [-] on Build 66
GenBankmRNA BY274561.1 - 2 - 3 227 Chr. 2 - 26637331 - 26640241 [-] on Build GRCm38 66
GenBankmRNA BY642400.1 - 1 - 2 48 Chr. 2 - 26637177 - 26639476 [-] on Build GRCm38 66
GenBankmRNA CA466784.1 - 2 - 3 267 Chr. 2 - 26637217 - 26640244 [-] on Build GRCm38 66
GenBankmRNA AA982539.1 - 2 - 3 202 Chr. 2 - 26637475 - 26640210 [-] on Build GRCm38 66
GenBankmRNA AV572272.1 - 2 - 3 215 Chr. 2 - 26637440 - 26640229 [-] on Build GRCm38 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small nucleolar RNA, H/ACA box 43 Snora43 ACA43 -56.0 Sense Chr. - 26637847 - 26637985 on Build GRCm38
Downstream small nucleolar RNA host gene 7, opposite strand Snhg7os 5730588L14Rik 902.0 Anti-sense Chr. - 26640373 - 26645944 on Build GRCm38

View Full Product Details