Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Has2os
- Entrez Gene ID
-
594843
- Gene Name
- hyaluronan synthase 2, opposite strand
- Gene Aliases
-
EG594843, Has2as, Hasnt
- Chromosome Location
-
Chr.15: 56689944 - 56694460 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002874.2 |
|
1 - 2 |
114 |
87 |
GenBank mRNA |
AY972478.1 |
- |
1 - 2 |
114 |
87 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
syntrophin, basic 1 |
Sntb1 |
|
-782.6kb
|
Anti-sense |
Chr. - 55638505 - 55907305 on Build GRCm38 |
Downstream |
solute carrier family 22 (organic cation transporter), member 22 |
Slc22a22 |
oat-pg |
550.0kb
|
Anti-sense |
Chr. - 57243767 - 57477625 on Build GRCm38 |
View Full Product Details