The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177632.3 | NP_808300.1 | 1 - 1 | 2050 | 107 |
GenBank mRNA | AK083036.1 | - | 1 - 1 | 2050 | 107 |
AK137950.1 | - | 1 - 1 | 2051 | 107 | |
BC022623.1 | - | 1 - 1 | 643 | 107 | |
BC026202.1 | - | 2 - 2 | 260 | 107 | |
BC099947.1 | - | 1 - 1 | 2063 | 107 | |
BC138882.1 | - | 1 - 1 | 1664 | 107 | |
BC138884.1 | - | 1 - 1 | 1640 | 107 |